Tisk…

Glycine 7-amido-4-methylcoumarin hydrobromide salt

Dodavatel: Apollo Scientific

BIB1440-100MG BIB1440-50MG
APOSBIB1440-100MGEA 5590 CZK
APOSBIB1440-100MG APOSBIB1440-50MG
Glycine 7-amido-4-methylcoumarin hydrobromide salt
Glycine 7-amido-4-methylcoumarin hydrobromide salt
Vzorec: C₁₂H₁₂N₂O₃·BrH
Storage Temperature: Ambient
MDL Number: MFCD00058452
CAS číslo: 113728-13-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR