Tisk…

Sulphosuccinimido biotin

Dodavatel: Apollo Scientific
Varování

Synonyms: Sulpho NHS biotin

BIB102-100MG
APOSBIB102-100MGEA 6220 CZK
APOSBIB102-100MG
Sulphosuccinimido biotin
Sulphosuccinimido biotin

Ref: LaRochelle, W.J. & Froehner, S.C.(1986)J.Biol.Chem.261 5270-5274Ref: Lee, W.T and Conrad, D.H. (1984) J.ExpMed 159, 1790-1795

Vzorec: C₁₄H₁₈N₃O₈S₂·Na
MW: 443.428 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00078535
CAS číslo: 194041-65-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR