Tisk…

Zebularine

Dodavatel: Apollo Scientific
Varování

Synonyms: 4-Deoxyuridine, 1-((2R,3R,4S,5R)-3,4-Dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidin-2(1H)-one

BIA8416-100MG BIA8416-50MG
APOSBIA8416-100MGEA 4570 CZK
APOSBIA8416-100MG APOSBIA8416-50MG
Zebularine
Zebularine
Vzorec: C₉H₁₂N₂O₅
Storage Temperature: Refrigerator
MDL Number: MFCD04973699
CAS číslo: 3690-10-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR