Tisk…
Decontaminant, DNA-ExitusPlus™ Indicator Free
Dodavatel: Apollo Scientific
BIA7409-5X500
APOSBIA7409-5X500EA 12900 CZK
APOSBIA7409-5X500
Decontaminant, DNA-ExitusPlus™ Indicator Free
Prostředky čistící Dekontaminanty
Indicator-free version of DNA-ExitusPlus™. Decontamination solution for the removal of DNA and RNA contaminations from surfaces.
Informace o dodávce: Supplied in spray bottles.