Tisk…

N-Succinimidyl Acetate

Dodavatel: Apollo Scientific
Varování

Synonyms: N-Acetoxysuccinimide, Ac-Osu, Acetic Acid N-Succinimidyl Ester, 2,5-Dioxopyrrolidin-1-yl acetate

BIA1015-10G BIA1015-25G BIA1015-100G
APOSBIA1015-10GEA 2070 CZK
APOSBIA1015-10G APOSBIA1015-25G APOSBIA1015-100G
N-Succinimidyl Acetate
N-Succinimidyl Acetate
Vzorec: C₆H₇NO₄
MW: 157.13 g/mol
Bod tání: 134 °C
Storage Temperature: Freezer
MDL Number: MFCD00037916
CAS číslo: 14464-29-0
EINECS: 238-454-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR