Tisk…

Potassium-L-aspartate

Dodavatel: Apollo Scientific
Varování

BIA0867-1KG
APOSBIA0867-1KGEA 1870 CZK
APOSBIA0867-1KG
Potassium-L-aspartate
Potassium-L-aspartate
Vzorec: C₄H₆NO₄K
Storage Temperature: Ambient
MDL Number: MFCD00021083
CAS číslo: 1115-63-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR