Tisk…

HEPES sodium salt ≥96%

Dodavatel: Apollo Scientific
Varování

BI6856-100G BI6856-25G BI6856-500G
APOSBI6856-100GEA 1160 CZK
APOSBI6856-100G APOSBI6856-25G APOSBI6856-500G
HEPES sodium salt ≥96%
HEPES sodium salt

Biological buffer.


  • Assay 96+%
  • Water (By Karl Fischer) < 3.0%
  • pH (1%) = 10.0 +/- 0.5

Vzorec: C₈H₁₇N₂NaO₄S
Storage Temperature: Ambient
MDL Number: MFCD00036463
CAS číslo: 75277-39-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR