Tisk…

5-Bromo-2'-deoxyuridine (broxuridine)

Dodavatel: Apollo Scientific
Nebezpečí

BI2048-1G BI2048-5G BI2048-25G
APOSBI2048-1GEA 705 CZK
APOSBI2048-1G APOSBI2048-5G APOSBI2048-25G
5-Bromo-2'-deoxyuridine (broxuridine)
5-Bromo-2'-deoxyuridine (broxuridine)

Thymidine analogue used as a mutagen in genetic research.

Vzorec: C₉H₁₁BrN₂O₅
Storage Temperature: Refrigerator
MDL Number: MFCD00006529
CAS číslo: 59-14-3
EINECS: 200-415-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR