Tisk…

Anti-Eph receptor A2+A3+A4 Rabbit Polyclonal Antibody

Dodavatel: ANTIBODIES.COM
A99112-100
ANTIA99112-100EA 13540 CZK
ANTIA99112-100
Anti-Eph receptor A2+A3+A4 Rabbit Polyclonal Antibody
Protilátky
Rabbit polyclonal antibody to EphA2 for WB and ELISA with samples derived from human and mouse.

  • Validated applications: WB, ELISA
  • Recommended dilutions: WB: 1:500 to 1:1000, ELISA: 1:10000

Type: Primary
Antigen: Eph receptor A2+A3+A4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
New Product
Objednat

Produktová specifikace

Symbol antigenu Eph receptor A2+A3+A4
Název antigenu ECK
Environmentally Preferable
Synonyma antigenu EPHA2|EPHA3|HEK|EPH Receptor A3|ETK1|EPHA4|EPH Receptor A4|Eph receptor A2+A3+A4|EPH Receptor A2
Druh protilátky Primární
Klonovalita Polyclonal
Spojení Unconjugated
Reaktivita Human,Mouse
Hostitel Rabbit
Identifikační číslo genu UniprotID# P29317
Izotyp IgG
Western blot Yes
ELISA Yes
Imunogen Synthetic peptide derived from human EphA2 (amino acids 533-582)
Formulář Liquid
Molekulární hmotnost 108 kDa
Čištění Purified from rabbit serum by antigen affinity chromatography using the immunising peptide
Skladovací pufr Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7,4, with 150 mM NaCl, 0,02% Sodium Azide, and 50% Glycerol
Skladovací teplota Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
Transportní teplota Shipped on blue ice at 4 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR