Tisk…

Bovine Histone H3 ELISA Kit

Dodavatel: ANTIBODIES.COM

A86997-96
ANTIA86997-96EA 19950 CZK
ANTIA86997-96
Bovine Histone H3 ELISA Kit
Test ELISY

Bovine Histone H3 ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of bovine Histone H3 in serum, plasma, tissue homogenates, and other biological fluids.


  • Ready-to-use ELISA Kit
  • Detection range: 31,25 to 2000 pg/ml
  • Sensitivity: 18,75 pg/ml
  • Sample type: Serum, plasma, tissue homogenates, and other biological fluids
  • Assay precision: Intra-Assay: CV <8%, Inter-Assay: CV <10%


Bovine Histone H3 ELISA kit employs the sandwich enzyme immunoassay technique for the quantitative measurement of bovine Histone H3 in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for Histone H3 has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Histone H3 present in each sample is bound to the wells by the immobilised antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-Histone H3 Antibody, which binds the captured Histone H3 present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualise the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of Histone H3 captured in each well. The concentration of Histone H3 can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.


Informace pro objednávání: This is a ready-to-use ELISA kit that contains: Pre-coated 96 well microplate, lyophilised standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin Conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR