Tisk…

Anti-PAP Rabbit Recombinant Antibody [clone: ACPP/8409R]

Dodavatel: ANTIBODIES.COM
A316844-100
ANTIA316844-100EA 19380 CZK
ANTIA316844-100
Anti-PAP Rabbit Recombinant Antibody [clone: ACPP/8409R]
Protilátky
Recombinant rabbit monoclonal [ACPP/8409R] antibody to PAP for IHC-P with samples derived from human.

  • Validated applications: IHC-P
  • Recommended dilutions: IHC: 1 to 2 µg/ ml

Type: Primary
Antigen: PAP
Clonality: Recombinant
Clone: ACPP/8409R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG kappa
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR