Tisk…

Human SLC14A1/UTE ELISA Kit

Dodavatel: ANTIBODIES.COM

A310073-96
ANTIA310073-96EA 15540 CZK
ANTIA310073-96
Human SLC14A1/UTE ELISA Kit
Test ELISY

Human SLC14A1/UTE ELISA kit is a 90 minutes sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human SLC14A1/UTE in serum, plasma, and other biological fluids.


  • Higher throughput: Get results in just 90 minutes, with a single wash step
  • Detection range: 1,25 to 20 ng/ml
  • Sensitivity: 0,043 ng/ml
  • Sample type: Serum, plasma or other biological fluids
  • Assay precision: Intra assay: CV <8%, Inter assay: CV <10%


Human SLC14A1/UTE ELISA kit (A310073) employs the sandwich enzyme immunoassay technique for the quantitative measurement of human SLC14A1/UTE in serum, plasma or other biological fluids. An antibody specific for SLC14A1/UTE has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the SLC14A1/UTE present in each sample is bound to the wells by the immobilised antibody. Biotinylated Anti-SLC14A1/UTE antibody, which also binds the SLC14A1/UTE present in each sample, and Streptavidin-HRP, which binds the Biotinylated Anti-SLC14A1/UTE antibody, are added and the microtiter plate is incubated. Following incubation, unbound Biotinylated Anti-SLC14A1/UTE antibody and unbound Streptavidin-HRP are removed by washing, and two substrate solutions are added to the wells. colour develops in proportion to the amount of SLC14A1/UTE captured in each well. The colour development is stopped by addition of stop solution which changes the colour from blue to yellow and the intensity of the colour is then measured. The concentration of SLC14A1/UTE in the samples can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.


Informace pro objednávání: This is a ready to use ELISA kit that contains: Pre-coated 96 well microplate, standard solution, standard diluent, biotinylated detection antibody, streptavidin-HRP, wash buffer (25X), substrate solution A, substrate solution B, stop solution, plate sealers, foil pouch, and protocol manual.

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR