Tisk…

Anti-HCN2 Mouse Monoclonal Antibody [clone: S71]

Dodavatel: ANTIBODIES.COM
A304767-100
ANTIA304767-100EA 13540 CZK
ANTIA304767-100
Anti-HCN2 Mouse Monoclonal Antibody [clone: S71]
Protilátky
Mouse monoclonal [S71] antibody to HCN2 for WB, IHC, ICC/IF, IP and antibody microarray with samples derived from human, mouse and rat.

  • Validated applications: WB, IHC, ICC/IF, IP, Antibody microarray
  • Recommended dilutions: WB: 1:1000, IHC: 1:1000, ICC/IF: 1:100

Type: Primary
Antigen: HCN2
Clonality: Monoclonal
Clone: S71
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human;Mouse;Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR