Tisk…

Anti-NCOR2/SMRT Rabbit Polyclonal Antibody

Dodavatel: ANTIBODIES.COM
A10415-100
ANTIA10415-100EA 11830 CZK
ANTIA10415-100
Anti-NCOR2/SMRT Rabbit Polyclonal Antibody
Protilátky
Rabbit polyclonal antibody to NCOR2 / SMRT for WB, IHC and ICC/IF with samples derived from human, mouse and rat.

  • Validated applications: WB, IHC, ICC/IF
  • Recommended dilutions: WB: 1:500 to 1:2000, IHC: 1:50 to 1:200, ICC/IF: 1:50 to 1:100

Type: Primary
Antigen: NCOR2/SMRT
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR