Tisk…
Examination gloves, nitrile, Micro-Touch® Nitrile
Dodavatel: ANSELL MEDICAL PRODUCTS
700151 700152 700153 700154 700155
ANSM700151EA 612 CZK
ANSM700151 ANSM700152 ANSM700153 ANSM700154 ANSM700155
Examination gloves, nitrile, Micro-Touch® Nitrile
Rukavice
These examination gloves provide an effective barrier against punctures and a wide range of chemicals while the white colour reveals soiling and contamination.
  • Free of natural rubber latex and powder, minimising the risk of Type I allergies
  • 'Thermo-elastic' material provides latex-like comfort and adapts to the hand's shape
  • Inner polymer coating facilitates donning, while the lack of powder considerably reduces the risk of skin abrasion
  • Micro-textured finger surfaces ensure a good grip on instruments
  • Economical 150-count packaging with 50% more gloves per
More Product Information