Tisk…
Encapsulated chemical protective suits for PAPR systems, AlphaTec® MICROGARD 3000, model 700
Dodavatel: Ansell
YE30T-00700-06 YE30T-00700-07 YE30T-00700-02 YE30T-00700-03 YE30T-00700-04 YE30T-00700-05
ANSEYE30T-00700-06EA 17020 CZK
ANSEYE30T-00700-06 ANSEYE30T-00700-07 ANSEYE30T-00700-02 ANSEYE30T-00700-03 ANSEYE30T-00700-04 ANSEYE30T-00700-05
Encapsulated chemical protective suits for PAPR systems, AlphaTec® MICROGARD 3000, model 700
Pracovní oděvy
Encapsulated chemical protective suits for use with PAPR systems.
  • Exceptional protection, with over 200 chemicals permeation tested, including chemical warfare agents
  • Welded and taped seams provide the highest barrier to liquids and particulates
  • Panoramic visor design ensures a good field of vision for the wearer
  • Double elasticated cuffs for a liquid-tight connection with chemical protective gloves
  • Air permeable SMS collar maintains sufficient air in the breathing zone while allowing excess air to flow into the suit body
  • Four exhalation valves allow a full
More Product Information
Objednat
Produktová specifikace
Environmentally Preferable
Aplikace oblečení
Risk area
Rod