Tisk…
Chemical resistant suit, Microchem® by AlphaTec® 4000, model 151-G02
Dodavatel: Ansell
GR40T-00151-03 GR40T-00151-04 GR40T-00151-05 GR40T-00151-06 GR40T-00151-07
ANSEGR40T-00151-03EA 32870 CZK
ANSEGR40T-00151-03 ANSEGR40T-00151-04 ANSEGR40T-00151-05 ANSEGR40T-00151-06 ANSEGR40T-00151-07
Chemical resistant suit, Microchem® by AlphaTec® 4000, model 151-G02
Jednorázové oděvy
Rear entry suit with neoprene rubber face seal for a close fit to full face respirators. Ideal for use in hazardous areas where protection against concentrated chemicals and biological agents is required.
  • Neoprene rubber face seal
  • Rear horizontal zip entry
  • Attached socks with boot overflap
  • Attached Ansell Barrier gloves, with oversleeves and finger loops
  • Ultrasonically welded and taped seams provide a high barrier to liquid and particulates
Typical applications include: Chemicals, oils and petrochemicals, pharmaceutical, agriculture, sewageMore Product Information
Objednat
Produktová specifikace
Model 151
Environmentally Preferable
Antistatický +
Rod
Aplikace oblečení Chemical protection,Laboratory,Industrial