Tisk…
Hydrometers, Anti-freeze Tester
Dodavatel: Amarell
H851710
AMARH851710EA 2330 CZK
AMARH851710
Hydrometers, Anti-freeze Tester
Hydrometry
In this hydrometer in glass tube with suction ball and plastic suction.
  • Paper scale with coloured indication the charge of indication °C status
  • Shot weighted body
  • Complete in cardboard box or in blister packing