Tisk…
Electronic precision thermometer for foodstuff
Dodavatel: Amarell
E910600
AMARE910600EA 0 CZK
AMARE910600
Electronic precision thermometer for foodstuff
Teploměry Teploměry digitální
Digital thermometer type ad 17th is a piercing thermometer, very well suitable for core measurements on roasting and freezing.
  • Wide measuring range
  • Precise PT100 probe of stainless steel
  • Handle of probe and PTFE cable resistant up to 250 °C
Upozornení: or 0,5 % of range