Tisk…
VOLCANO Multirisk Trousers
Dodavatel: ALSICO
1030939-BE38 1030939-BE40 1030939-BE42 1030939-BE44 1030939-BE46 1030939-BE48 1030939-BE50 1030939-BE52 1030939-BE54 1030939-BE56 1030939-BE58 1030939-BE60 1030939-BE62 1030939-BE64 1030940-BE38 1030940-BE40 1030940-BE42 1030940-BE44 1030940-BE46 1030940-BE48 1030940-BE50 1030940-BE52 1030940-BE54 1030940-BE56 1030940-BE58 1030940-BE60 1030940-BE62 1030940-BE64
ALSW1030939-BE38EA 0 CZK
ALSW1030939-BE38 ALSW1030939-BE40 ALSW1030939-BE42 ALSW1030939-BE44 ALSW1030939-BE46 ALSW1030939-BE48 ALSW1030939-BE50 ALSW1030939-BE52 ALSW1030939-BE54 ALSW1030939-BE56 ALSW1030939-BE58 ALSW1030939-BE60 ALSW1030939-BE62 ALSW1030939-BE64 ALSW1030940-BE38 ALSW1030940-BE40 ALSW1030940-BE42 ALSW1030940-BE44 ALSW1030940-BE46 ALSW1030940-BE48 ALSW1030940-BE50 ALSW1030940-BE52 ALSW1030940-BE54 ALSW1030940-BE56 ALSW1030940-BE58 ALSW1030940-BE60 ALSW1030940-BE62 ALSW1030940-BE64
VOLCANO Multirisk Trousers
Pracovní oděvy
Comfortable multirisk trousers in inherent flame retardant fabric.
  • Preformed, modern fit
  • Elasticated waistband with button hole
  • Closure with jeans button and zipped fly
  • 2 large, straight inserted side pockets
  • Thigh pocket with expansion slit and phone pocket, closed with flap and hidden press studs
  • Double metre pocket with flap
  • 2 large back pockets with expansion slit, reinforced and closed with flap and hidden press studs
  • Preformed, closable knee pockets in main fabric
  • Extendable trouser legs
  • Piping in accent colour
Industrial launderable.