Tisk…
Sodium Standard Solution, 10000 mg/L Na in dil. Nitric Acid Standard for ICP-MS
Dodavatel: ACTIVE MOTIF
M135.2NP.L1
ACSDM135.2NP.L1EA 0 CZK
ACSDM135.2NP.L1
Sodium Standard Solution, 10000 mg/L Na in dil. Nitric Acid Standard for ICP-MS
Standardy Standardy kationické Standardy - sodík
Na 10000 mg/L in diluted nitric acid for ICP/MS.
Shelf life on ship date: 36 months - store under normal laboratory conditions, at temperatures between 15 to 25 °C.
Certifikace: ISO 17034, ISO 17025, ISO 9001.