Tisk…

Nalidixic acid sodium salt

Dodavatel: Thermo Fisher Scientific
Nebezpečí

463550010 463550050 463550250
ACRO463550010EA 986 CZK
ACRO463550010 ACRO463550050 ACRO463550250
Nalidixic acid sodium salt
Nalidixic acid sodium salt

  • Bioactive

Vzorec: C₁₂H₁₁N₂NaO₃
Bod tání: >280 °C
Storage Temperature: Ambient
MDL Number: MFCD00064376
CAS číslo: 3374-05-8

Objednat

Výsledky specifikačního testu

Appearance White to light yellow Powder
Infrared spectrum Conforms
Water ≤7 %
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR