Tisk…

MOPSO sodium salt (3-(4-morpholino)-2-hydroxypropanesulphonic acid sodium salt)

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: Sodium 2-hydroxy-3-morpholinopropane-1-sulphonate

462140250 462141000 462145000
ACRO462140250EA 2450 CZK
ACRO462140250 ACRO462141000 ACRO462145000
MOPSO sodium salt (3-(4-morpholino)-2-hydroxypropanesulphonic acid sodium salt)
MOPSO sodium salt (3-(4-morpholino)-2-hydroxypropanesulphonic acid sodium salt)

  • Bioactive

Vzorec: C₇H₁₄NNaO₅S
Storage Temperature: Ambient
MDL Number: MFCD00070007
CAS číslo: 79803-73-9

Objednat

Výsledky specifikačního testu

Appearance White Crystalline powder
Infrared spectrum Conforms
Titration Total base ≥98.5 %
Water ≤0.5 %
UV at 290 nm A: ≤0.1 (33% W/W)
Heavy metals (as Pb) ≤5 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR