Tisk…

Fluticasone propionate

Dodavatel: Thermo Fisher Scientific

462101000 462105000
ACRO462101000EA 1960 CZK
ACRO462101000 ACRO462105000
Fluticasone propionate
Fluticasone propionate
Vzorec: C₂₅H₃₁F₃O₅S
Bod tání: 272…273 °C
Storage Temperature: Freezer
MDL Number: MFCD00866007
CAS číslo: 80474-14-2

Objednat

Výsledky specifikačního testu

Appearance White to off-white Powder
Infrared spectrum Conforms
Loss on drying ≤0.5 %
HPLC ≥96.0 %
Specific optical rotation +32° to +36° (20°C, 589 nm) (0.5% in CH2Cl2)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR