Tisk…

Carbidopa monohydrate 99%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: (S)-3-(3,4-Dihydroxyphenyl)-2-hydrazino-2-methylpropionic Acid

461010010
ACRO461010010EA 5890 CZK
ACRO461010010
Carbidopa monohydrate 99%
Carbidopa monohydrate
Vzorec: C₁₀H₁₄N₂O₄
MW: 226.23 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00889211
CAS číslo: 38821-49-7

Objednat

Výsledky specifikačního testu

Appearance White to yellowish-white Powder
Infrared spectrum Conforms
Loss on drying 6.9 % to 7.9 % (1 g, 105°C)
HPLC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR