Tisk…

Methyl myristate 99% analytical standard for GC

Dodavatel: Thermo Fisher Scientific

460360010 460360050
ACRO460360010EA 1480 CZK
ACRO460360010 ACRO460360050
Methyl myristate 99% analytical standard for GC
Methyl myristate
Vzorec: C₁₅H₃₀O₂
Bod varu: 323 °C (760 mmHg)
Hustota: 0,863 g/cm³
Bod vzplanutí: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00008983
CAS číslo: 124-10-7

Objednat

Výsledky specifikačního testu

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4360 to 1.4380 (20°C, 589 nm)
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR