Tisk…

Florisil® 30-60 mesh for column chromatography

Dodavatel: Thermo Fisher Scientific

Synonyms: Magnesium silicate, aktivní, Florisil

457735000
ACRO457735000EA 7800 CZK
ACRO457735000
Florisil® 30-60 mesh for column chromatography
Magnesium silicate
Vzorec: MgO₃Si
MW: 100.38 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00078249
CAS číslo: 1343-88-0

Objednat

Výsledky specifikačního testu

Appearance White Powder (May contain black particles)
Silicate (as SiO2) 83.5 to 84.5 %
Sodium sulfate (Na2SO4) ≤1.0
Magnesium oxide (MgO) 15 to 16 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR