Tisk…

Thiamphenicol 98%

Dodavatel: Thermo Fisher Scientific

455450010 455450050 455450250
ACRO455450010EA 751 CZK
ACRO455450010 ACRO455450050 ACRO455450250
Thiamphenicol 98%
Thiamphenicol
Vzorec: C₁₂H₁₅Cl₂NO₅S
Storage Temperature: Ambient
MDL Number: MFCD00467983
CAS číslo: 15318-45-3

Objednat

Výsledky specifikačního testu

Appearance White to off-white Powder
Infrared spectrum Conforms
Melting point 160 to 170 °C
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR