Tisk…

N(α)-Acetyl-L-tryptophan 97%

Dodavatel: Thermo Fisher Scientific

451650050 451650250 451651000
ACRO451650050EA 1160 CZK
ACRO451650050 ACRO451650250 ACRO451651000
N(α)-Acetyl-L-tryptophan 97%
N(α)-Acetyl-L-tryptophan
Vzorec: C₁₃H₁₄N₂O₃
Bod tání: 186…192 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00065976
CAS číslo: 1218-34-4

Objednat

Výsledky specifikačního testu

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Assay ≥96.0 %
Loss on drying ≤0.3 %
Specific optical rotation +23° to +28° (20°C, 589 nm) (c=1, EtOH)

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR