Tisk…

Bentonit

Dodavatel: Thermo Fisher Scientific

Synonyms: Montmorillonite, Bentonite

447440025 447445000
ACRO447440025EA 3760 CZK
ACRO447440025 ACRO447445000
Bentonit
Bentonite
Storage Temperature: Ambient
MDL Number: MFCD00130611
CAS číslo: 1302-78-9

Objednat

Výsledky specifikačního testu

Appearance White to grey to brown Crystalline powder and/or chunks

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR