Tisk…

Dihydrogenfosforečnan amonný 99% for HPLC

Dodavatel: Thermo Fisher Scientific

Synonyms: Ammonium phosphate, monobasic, Ammonium Phosphate Monobasic, Ammonium dihydrogenphosphate

447382500
ACRO447382500EA 9010 CZK
ACRO447382500
Dihydrogenfosforečnan amonný 99% for HPLC
Dihydrogenfosforečnan amonný
Vzorec: NH₄H₂PO₄
Bod tání: 190 °C
Hustota: 1,8 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003396
CAS číslo: 7722-76-1

Objednat

Výsledky specifikačního testu

Appearance White Crystalline powder or crystals
Water ≤1.0 %
Solubility (10 % in water) Clear colorless
UV at 280 nm A: ≤0.05
pH 4.0 to 5.0 (10 % in water)
Titration with NaOH ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR