Tisk…

Gallium(III) chloride, anhydrous 99.99+% (trace metals basis), beads -10 mesh

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: Gallium trichloride

444100050 444100250 444101000
ACRO444100050EA 3070 CZK
ACRO444100050 ACRO444100250 ACRO444101000
Gallium(III) chloride, anhydrous 99.99+% (trace metals basis), beads -10 mesh
Gallium(III) chloride
Vzorec: Cl₃Ga
Bod varu: 201 °C (760 mmHg)
Bod tání: 78 °C
Hustota: 2,29 g/cm³ (80 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011018
CAS číslo: 13450-90-3
UN: 3260
ADR: 8,II

Objednat

Výsledky specifikačního testu

Appearance White to off-white Beads
Total trace metal impurities ≤100 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR