Tisk…

Sodium methanesulphonate 99%

Dodavatel: Thermo Fisher Scientific

442110050 442111000 442115000
ACRO442110050EA 1310 CZK
ACRO442110050 ACRO442111000 ACRO442115000
Sodium methanesulphonate 99%
Sodium methanesulphonate
Vzorec: CH₃NaO₃S
Storage Temperature: Ambient
MDL Number: MFCD00064389
CAS číslo: 2386-57-4

Objednat

Výsledky specifikačního testu

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Water ≤1 %
Titration after Ion exchange ≥98.5 %
Free acid ≤1 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR