Tisk…

Nortropinone hydrochloride 97%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: 8-Azabicyclo[3.2.1]octan-3-one hydrochloride

436120010 436120050
ACRO436120010EA 2920 CZK
ACRO436120010 ACRO436120050
Nortropinone hydrochloride 97%
Nortropinone hydrochloride
Vzorec: C₇H₁₂ClNO
MW: 161.63 g/mol
Bod tání: 194.7…195.8 °C (rozkl.)
Storage Temperature: Ambient
MDL Number: MFCD03613582
CAS číslo: 25602-68-0

Objednat

Výsledky specifikačního testu

Appearance White to off-white to light brown Powder or solid
Infrared spectrum Conforms
Titration Argentometric ≥96.0 to ≤104.0 %
GC ≥98.0 %
Specific optical rotation +12° to +15° (20°C, 589 nm) (Neat)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR