Tisk…

Imidazo[1,2-a]pyridine-2-carboxylic acid hydrate 97%

Dodavatel: Thermo Fisher Scientific
Varování

432780010 432780050
ACRO432780010EA 2960 CZK
ACRO432780010 ACRO432780050
Imidazo[1,2-a]pyridine-2-carboxylic acid hydrate 97%
Imidazo[1,2-a]pyridine-2-carboxylic acid hydrate
Vzorec: C₈H₆N₂O₂·xH₂O
Bod tání: 232…234 °C
Storage Temperature: Ambient
MDL Number: MFCD17171427
CAS číslo: 64951-08-2

Objednat

Výsledky specifikačního testu

Appearance White to off-white Powder
Infrared spectrum Conforms
Additional info Hydrate, contains 9.5% to 11.0% water
HPLC ≥96.0 %
Elemental Analysis 14.96 to 16.17 % N
Residue after evaporation ≤0.01 %
Peroxides ≤50 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR