Tisk…

N-Succinimidyl Bromoacetate 90%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: 2,5-Dioxopyrrolidin-1-yl 2-bromoacetate, N-(Bromoacetoxy)succinimide, Bromoacetic Acid N-Succinimidyl Ester, Bromoacetic acid N-hydroxysuccinimide ester

427780010 427780050 427782500
ACRO427780010EA 29930 CZK
ACRO427780010 ACRO427780050 ACRO427782500
N-Succinimidyl Bromoacetate 90%
N-Succinimidyl Bromoacetate
Vzorec: C₆H₆BrNO₄
MW: 236.02 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00058571
CAS číslo: 42014-51-7

Objednat

Výsledky specifikačního testu

Appearance White to off-white Powder
Infrared spectrum Conforms
Elemental Analysis ≥88 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR