Tisk…

2-(Diethylamino)ethyl bromide hydrobromide 98%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

426860250 426861000
ACRO426860250EA 3900 CZK
ACRO426860250 ACRO426861000
2-(Diethylamino)ethyl bromide hydrobromide 98%
2-(Diethylamino)ethyl bromide hydrobromide

Appearance: White to yellow Powder or solid

Vzorec: C₆H₁₄BrN
Bod tání: 211…215 °C
Storage Temperature: Ambient
MDL Number: MFCD00040376
CAS číslo: 1069-72-3
UN: 2811
ADR: 6.1,III

Objednat

Výsledky specifikačního testu

Appearance White to yellow Powder or solid
Infrared spectrum Conforms
Titration Argentometric ≥97.5 %
NMR (1H-NMR) Consistent with structure


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR