Tisk…

N-(tert-Butoxycarbonyl)-1,4-diaminobutane 95%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

426190010 426190050
ACRO426190010EA 2390 CZK
ACRO426190010 ACRO426190050
N-(tert-Butoxycarbonyl)-1,4-diaminobutane 95%
N-(tert-Butoxycarbonyl)-1,4-diaminobutane

Appearance: Clear colorless to light yellow Liquid

Vzorec: C₉H₂₀N₂O₂
Hustota: 0,984 g/cm³
Bod vzplanutí: 109 °C
Storage Temperature: Ambient
MDL Number: MFCD00210019
CAS číslo: 68076-36-8
UN: 2735
ADR: 8,III

Objednat

Výsledky specifikačního testu

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
GC ≥94 %
NMR consistent with structure


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR