Tisk…

Močovina 99% ACS

Dodavatel: Thermo Fisher Scientific

Synonyms: Carbonyl diamide, Carbamide, Diaminomethanone, Carbonyldiamide

424585000 424580025 424580100 424581000
ACRO424585000EA 1910 CZK
ACRO424585000 ACRO424580025 ACRO424580100 ACRO424581000
Močovina 99% ACS
Močovina
Vzorec: NH₂CONH₂
MW: 60.06
Bod varu: 197
Bod tání: 131…135 °C
Storage Temperature: Ambient
MDL Number: MFCD00008022
CAS číslo: 57-13-6
EINECS: 200-315-5
REACH: 01-2119463277-33
Merck Index: 13,09935
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a9682f71b212a449398841e2beace2d7500b4710&vid=d6057a7ec3d606031d338a300b5d5db30b52d861&prid=web&clid=SAPDAM

Objednat

Výsledky specifikačního testu

Appearance White Crystals
Infrared spectrum Conforms
Melting point 132 to 135 °C
Residue after ignition ≤0.01 %
Titration with HClO4 99.0 to 100.5 %
Chloride (Cl) ≤5 ppm
Sulfate (SO4) ≤10 ppm
Iron (Fe) ≤10 ppm
Heavy metals (as Pb) ≤10 ppm
Insoluble matter ≤0.01 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR