Tisk…

N-2-Aminoethyl-1-naftylamin dihydrochlorid 98+% ACS

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: N-2-Aminoethyl-1-naftylamin dihydrochlorid

423990250 423991000 423995000 423990050
ACRO423990250EA 6550 CZK
ACRO423990250 ACRO423991000 ACRO423995000 ACRO423990050
N-2-Aminoethyl-1-naftylamin dihydrochlorid 98+% ACS
N-(1-Naphthyl)ethylenediamine dihydrochlorid
Vzorec: C₁₂H₁₆Cl₂N₂
Storage Temperature: Ambient
MDL Number: MFCD00012556
CAS číslo: 1465-25-4
EINECS: 215-981-2
Merck Index: 12,06496
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3643e6df3e9cf8dc627823e9090c97004ee25308&vid=a30fc28279c11af12c6b735b00b84fc8f08caf34&prid=web&clid=SAPDAM

Objednat

Výsledky specifikačního testu

Appearance White to beige to pink Powder or crystals and/or chunks
Infrared spectrum Conforms
Titration Argentometric ≥98.0 % (on dry substance)
Water ≤5 % (K.F.)
Loss on drying ≤5 % (1 g, 80°C, 4 h)
Solubility Passes test
Sensitivity (to sulfanilamide) Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR