Tisk…

Oxid vápenatý 97+%, powder for analysis

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: Quicklime, Calcium oxide from marble, caustic, Lime

422830010 422830025
ACRO422830010EA 1130 CZK
ACRO422830010 ACRO422830025
Oxid vápenatý 97+%, powder for analysis
Oxid vápenatý
Vzorec: CaO
Bod varu: 2850 °C (760 mmHg)
Bod tání: 2570 °C
Hustota: 3,38 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010911
CAS číslo: 1305-78-8
EINECS: 215-138-9
UN: 1910
ADR: 8,III
Merck Index: 13,01691

Objednat

Výsledky specifikačního testu

Appearance White to off-white to light yellow Powder
Titration Complexometric ≥97.0 % (CaO) on dry substance
Nitrate (NO3) ≤500 ppm
Sulfate (SO4) ≤1000 ppm
Iron (Fe) ≤1000 ppm
Heavy metals (as Pb) ≤100 ppm
Insoluble matter ≤1 % (in acetic acid)
NH4OH precipitate ≤2.0 %
Loss on ignition ≤5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR