Tisk…

Saponin (from Quillaja Saponaria Molina) pract.

Dodavatel: Thermo Fisher Scientific

419231000 419235000 419230025
ACRO419231000EA 3190 CZK
ACRO419231000 ACRO419235000 ACRO419230025
Saponin (from Quillaja Saponaria Molina) pract.
Saponin
Storage Temperature: Ambient
MDL Number: MFCD00081981
CAS číslo: 74499-23-3

Objednat

Výsledky specifikačního testu

Appearance Dark red to tan Powder
Infrared spectrum Conforms
Solubility (20 % in water) Clear to hazy foaming solution
Moisture ≤10 %
Ash ≤10 %
pH 3.8 to 4.6 (20 % solution)

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR