Tisk…

3-Methyl-2-benzothiazolinone hydrazone hydrochloride monohydrate 98%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: 2-Hydrazono-3-methyl-2,3-dihydrobenzo[d]thiazole hydrochloride hydrate, 2-Hydrazono-3-methylbenzothiazoline hydrochloride

414020100 414020500 414022500
ACRO414020100EA 3620 CZK
ACRO414020100 ACRO414020500 ACRO414022500
3-Methyl-2-benzothiazolinone hydrazone hydrochloride monohydrate 98%
3-Methyl-2-benzothiazolinone hydrazone hydrochloride monohydrate

Appearance: White to beige Crystalline powder

Vzorec: C₈H₉N₃S·H₂O·HCl
MW: 233.714 g/mol
Bod tání: 270…276 °C (rozkl.)
Storage Temperature: Ambient
MDL Number: MFCD00149370
CAS číslo: 38894-11-0
UN: 2811
ADR: 6.1 III

Objednat

Výsledky specifikačního testu

Appearance White to beige Crystalline powder
Infrared spectrum Conforms
Loss on drying ≤8 %
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR