Tisk…

D(+)-Glukóza, anhydrous ACS

Dodavatel: Thermo Fisher Scientific

Synonyms: Corn sugar, ᴅ-(+)-Glucose, Glucose, D-(+)-Dextrose, (2R,3S,4R,5R)-2,3,4,5,6-Pentahydroxyhexanal, Glucolin, Dextrose, D-Glucose

410950010 410955000 410950050
ACRO410950010EA 3020 CZK
ACRO410950010 ACRO410955000 ACRO410950050
D(+)-Glukóza, anhydrous ACS
D(+)-Glukóza
Vzorec: C₆H₁₂O₆
MW: 180.16
Bod tání: 146 °C
Storage Temperature: Ambient
MDL Number: MFCD00063684
CAS číslo: 50-99-7
EINECS: 200-075-1
Merck Index: 14,04459
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=47ec28c7fd4803097405f8eaef3171b3a40d40c5&vid=62627c96e2ae9f2a0cae732d0ddf0a100247298e&prid=web&clid=SAPDAM

Objednat

Výsledky specifikačního testu

Appearance White Crystalline powder
Infrared spectrum Conforms
Loss on drying ≤0.2 % (105°C, 6 h)
Residue after ignition ≤0.02 %
Specific optical rotation +52.5° to +53.0° (25°C, 589 nm) (c=10, H2O, NH4OH)
Chloride (Cl) ≤100 ppm
SO4 and SO3 (as SO4) ≤0.005 %
Iron (Fe) ≤5 ppm
Heavy metals (as Pb) ≤5 ppm
Titrable acid ≤0.002 meq/g
Insoluble matter ≤0.005 %
Starch Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR