Tisk…

Ethyl phenylglyoxylate 95%

Dodavatel: Thermo Fisher Scientific

410350050 410350250
ACRO410350050EA 861 CZK
ACRO410350050 ACRO410350250
Ethyl phenylglyoxylate 95%
Ethyl phenylglyoxylate
Vzorec: C₁₀H₁₀O₃
Bod varu: 138…139 °C (18 mmHg)
Bod vzplanutí: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00009120
CAS číslo: 1603-79-8
EINECS: 216-504-0

Objednat

Výsledky specifikačního testu

Appearance Clear colorless to green Liquid
Infrared spectrum Conforms
Refractive index 1.5150 to 1.5170 (20°C, 589 nm)
GC ≥94 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR