Tisk…

Diethylether 99 Stabilised HPLC

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: Ethyl Ether, Ether, 2,2'-Oxydiethane, 2,2'-Oxybisethane, Ethoxyethane

Stabilization: EtOH

410040010 410040025
ACRO410040010EA 2430 CZK
ACRO410040010 ACRO410040025
Diethylether 99 Stabilised HPLC
Diethylether
Vzorec: (CH₃CH₂)₂O
MW: 74.12
Bod varu: 35
Bod tání: –116 °C
Hustota: 0.714 g/cm³
Bod vzplanutí: –45 °C
Storage Temperature: Ambient
MDL Number: MFCD00011646
CAS číslo: 60-29-7
UN: 1155
ADR: 3 I
Merck Index: 12,03852
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=16f2debd1f18f9bff9952c4a6e5d599a67f94e61&vid=3dd6c545e58e36e1004e828026523fb0e3bcbbcd&prid=web&clid=SAPDAM

Objednat

Výsledky specifikačního testu

Appearance Clear liquid
Water ≤0.02 %
GC ≥99 %
UV at 280 nm A: ≤0.005
Color scale ≤10 APHA
Stabilizer 2 % Ethanol
Wavelength at 1 A.U. ≤205 nm
Residue after evaporation ≤2 ppm
Acidity/alkalinity ≤0.0002 meq/g


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR