Tisk…

DNA sodium salt (Deoxyribonucleic acid sodium salt), M.W. 50000-100000 Da, pure

Dodavatel: Thermo Fisher Scientific

Synonyms: Thymonucleic acid sodium salt, DNA sodium salt

406250250 406251000
ACRO406250250EA 2550 CZK
ACRO406250250 ACRO406251000
DNA sodium salt (Deoxyribonucleic acid sodium salt), M.W. 50000-100000 Da, pure
DNA sodium salt (Deoxyribonucleic acid sodium salt)

CAS No.: 68938-01-2


Appearance: Off-white to tan Powder

Storage Temperature: Ambient
MDL Number: MFCD00130922
CAS číslo: 68938-01-2

Objednat

Výsledky specifikačního testu

Appearance Almost white to light yellow to tan Powder
Infrared spectrum Conforms
Loss on drying ≤18 %
Protein ≤5 %
Phosphorus (P) ≤9 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR