Tisk…

Benzoylchlorid 98+% ACS

Dodavatel: Thermo Fisher Scientific
Nebezpečí

402030025 402030050 402035000
ACRO402030025EA 7650 CZK
ACRO402030025 ACRO402030050 ACRO402035000
Benzoylchlorid 98+% ACS
Benzoylchlorid
Vzorec: C₇H₅ClO
Bod varu: 197,2 °C (1013 hPa)
Bod tání: –1 °C
Hustota: 1,21 g/cm³
Bod vzplanutí: 93 °C
Storage Temperature: Ambient
MDL Number: MFCD00000653
CAS číslo: 98-88-4
EINECS: 202-710-8
UN: 1736
ADR: 8,II
Merck Index: 13,01113

Objednat

Výsledky specifikačního testu

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.5520 to 1.5540 (20°C, 589 nm)
Residue after ignition ≤0.005 %
Titration excess NaOH 98.0 to 100.5 %
Iron (Fe) ≤0.001 %
Heavy metals (as Pb) ≤10 ppm
Phosphorous compounds (as P) ≤0.002 %
Freezing point -2.0 to 0.0°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR