Tisk…

Acid orange 5 indicator grade

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: OrangeIV, Aniline yellow, 4-(4-Anilinophenylazo)benzenesulphonic acid sodium salt, CI 13080, Diphenylamine orange, Indicator, S. No.: 179, Orange IV, Tropaeolin OO

401261000 401260250 401260010
ACRO401261000EA 10410 CZK
ACRO401261000 ACRO401260250 ACRO401260010
Acid orange 5 indicator grade
Acid orange 5

CAS No.: 554-73-4

Vzorec: C₁₈H₁₄N₃NaO₃S
Storage Temperature: Ambient
MDL Number: MFCD00038130
CAS číslo: 554-73-4

Objednat

Výsledky specifikačního testu

Appearance Ochre to orange Powder
Infrared spectrum Conforms
Visual transition interval from pH 1.4 (red) to pH 2.6 (yellow)
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR