Tisk…

Peroxodisíran amonný 98+% ACS

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: Peroxidisulphuric acid diammonium salt, Ammonium persulphate

401160020 401160100 401161000 401165000 401160050 401160250
ACRO401160020EA 4230 CZK
ACRO401160020 ACRO401160100 ACRO401161000 ACRO401165000 ACRO401160050 ACRO401160250
Peroxodisíran amonný 98+% ACS
Peroxodisíran amonný
Vzorec: (NH₄)₂S₂O₈
Hustota: 1,98 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00003390
CAS číslo: 7727-54-0
EINECS: 231-786-5
UN: 1444
ADR: 5.1,III
Merck Index: 13,00545
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f82a373f21d34e63dc94c0598f3f33ba6bbfdb2b&vid=2b6d196dbf07ce4dd528ac7b4f8456678accd3ba&prid=web&clid=SAPDAM

Objednat

Výsledky specifikačního testu

Appearance White Crystalline powder or crystals
Residue after ignition ≤0.05 %
Titration with KMnO4 ≥98.0 %
Chloride and Chlorate (as Cl) ≤0.001 %
Iron (Fe) ≤0.001 %
Manganese (Mn) ≤0.5 ppm
Heavy metals (as Pb) ≤0.005 %
Titrable free acid ≤0.04 meq/g
Insoluble matter ≤0.005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR